missing translation for 'onlineSavingsMsg'
Learn More

Phenol, Saturated (pH 6.6/7.9, Liq.), Fisher BioReagents™

Product Code. 10001173
Change view
Click to view available options
Quantity:
100 mL
400 mL
Packaging:
Coated Amber Glass/Poison Pack
Unit Size:
100mL
400mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10001173 400 mL Coated Amber Glass/Poison Pack 400mL
10264673 100 mL Coated Amber Glass/Poison Pack 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10001173 Supplier Fisher BioReagents Supplier No. BP1750I400
 Restricted Item
Estimated Shipment: 26-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Phenol, Saturated (pH 6.6/7.9, Liq.)

Phenol saturated with Tris buffer is used in DNA extraction procedures.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 108-95-2 , 6381-92-6 , 77-86-1
Molecular Formula C6H6O
Molecular Weight (g/mol) 94.11
MDL Number MFCD00002143
InChI Key ISWSIDIOOBJBQZ-UHFFFAOYSA-N
Synonym carbolic acid, hydroxybenzene, phenic acid, phenylic acid, oxybenzene, benzenol, phenyl hydrate, monophenol, phenyl hydroxide, phenylic alcohol
PubChem CID 996
ChEBI CHEBI:15882
IUPAC Name phenol
SMILES OC1=CC=CC=C1

Specifications

Assay Percent Range ≥99.0
UN Number 2821
Percent Purity ≥99.0%
Identification Pass Test
Packaging Coated Amber Glass/Poison Pack
Quantity 400 mL
Physical Form Liquid
Chemical Name or Material Phenol, Saturated
Grade DNase- and RNase-Free
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • Phenol saturated pH 6.6
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Skin corrosion/irritation Category 1 B
  • Serious eye damage/eye irritation Category 1
  • Germ cell mutagenicity Category 2
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H301-Toxic if swallowed.
  • H311-Toxic in contact with skin.
  • H331-Toxic if inhaled.
  • H314-Causes severe skin burns and eye damage.
  • H341-Suspected of causing genetic defects.
  • H373-May cause damage to organs.
  • H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contain: Phenol, Ethylenediaminetetraacetic aciddisodium salt dihydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.