missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Phenol water saturated sol., for molecular biology, DNAse, RNAse, Protease free, stab., free of oxides

Product Code. 10458872 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
500 mL
Unit Size:
500mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10458872 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10458872 Supplier Thermo Scientific Chemicals Supplier No. 327105000
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

Phenol/TRIS , CAS # 96210-45-6, is a molecular biology solvent system that can be used for the isolation of DNA. | CAS: 108-95-2 | C6H6O | 94.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General description

• Phenol/Tris is a solvent system consisting of phenol and Tris(Hydroxymethyl)aminomethane
• Prepared as ready-to-use, no titration necessary

Applications

• Commonly used in DNA isolation. Tris(Hydroxymethyl)aminomethane acts as a proton acceptor to raise pH

TRUSTED_SUSTAINABILITY

Specifications

Color Colorless to Yellow
CAS 108-95-2
Molecular Formula C6H6O
Linear Formula OHC6H5
MDL Number MFCD00002143
Beilstein 06, 110
Fieser 01,828; 13,166; 14,88
Density 1.0600g/mL
Merck Index 15, 7354
Synonym carbolic acid, hydroxybenzene, phenic acid, phenylic acid, oxybenzene, benzenol, phenyl hydrate, monophenol, phenyl hydroxide, phenylic alcohol
InChI Key ISWSIDIOOBJBQZ-UHFFFAOYSA-N
SMILES OC1=CC=CC=C1
IUPAC Name phenol
Molecular Weight (g/mol) 94.11
PubChem CID 996
ChEBI CHEBI:15882
Formula Weight 94.11
Grade Molecular Biology
Specific Gravity 1.06
Quantity 500 mL
Flash Point 79°C
Physical Form Liquid
Chemical Name or Material Phenol water saturated sol., DNase, RNase, Protease Free, Stabilized, Free of Oxides
Show More Show Less
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • Phenol water saturated solution,DNase,RNase,Protease free,stab., free of oxides
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Serious eye damage/eye irritation Category 1
  • Germ cell mutagenicity Category 2
  • Skin corrosion/irritation Category 1 B
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H301-Toxic if swallowed.
  • H311-Toxic in contact with skin.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • H331-Toxic if inhaled.
  • H341-Suspected of causing genetic defects.
  • H373-May cause damage to organs.
  • H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contain: Phenol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.