missing translation for 'onlineSavingsMsg'
Learn More

Phenolics Mix A, SPEX CertiPrep™

ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

Brand:  SPEX Certiprep CLPS-AI

Additional Details : Weight : 0.00100kg Transport : UN number : 1219 Chem class : 3 Pack group : II


Product Code. 16339877

  • 109.00 € / 1mL
Estimated Shipment: 19-09-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Includes: Autosampler Vial, NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

Description

Description

Organic Standard manufactured for GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation

AnalyteCAS No.Concentration
2,4,6-Trichlorophenol88-06-22000μg/mL
2,4-Dichlorophenol120-83-22000μg/mL
2,4-Dimethylphenol105-67-92000μg/mL
2,4-Dinitrophenol51-28-52000μg/mL
2-Chlorophenol95-57-82000μg/mL
2-Nitrophenol88-75-52000μg/mL
4,6-Dinitro-2-methylphenol534-52-12000μg/mL
4-Chloro-3-methylphenol59-50-72000μg/mL
4-Nitrophenol100-02-72000μg/mL
Pentachlorophenol87-86-52000μg/mL
Phenol108-95-22000μg/mL

Certifications

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

Compliance

GHS/OSHA Complaint

Specifications

Specifications

Phenolics Mix A
2mL amber prescored glass ampule
Isopropanol
GC, GC/MS, HPLC, LC/MS, and Other Analytical Instrumentation
SW-846 Method 8040
For US EPA Method 8040
1 mL
Certificate of Analysis reports target analytes to a +/−2% accuracy and correct for raw material purity.
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Signal Word
  • Danger

Hazard Category
  • Carcinogenicity Category 2
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
  • Skin sensitiser Category 1
  • Specific target organ toxicity Category 3

Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H317-May cause an allergic skin reaction.
  • H319-Causes serious eye irritation.
  • H336-May cause drowsiness or dizziness.
  • H351-Suspected of causing cancer.

Precautionary Statement
  • P101-If medical advice is needed, have product container or label at hand.
  • P102-Keep out of reach of children.
  • P103-Read label before use.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

Supplemental information
  • MIXTURE LIST-Contains : Propan-2-ol, 2,4,6-trichlorophenol, 2-chlorophenol, DNOC, chlorocresol, pentachlorophenol, 2,4-dichl

SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product intended for laboratory use only