missing translation for 'onlineSavingsMsg'
Learn More

Phenothrin, Dr. Ehrenstorfer

Product Code. 30327096
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30327096 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30327096 Supplier Dr. Ehrenstorfer Supplier No. DREC16040000
 Restricted Item
Estimated Shipment: 01-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Phenothrin
CAS 26002-80-2
Analyte or Component Names Phenothrin
Shipping Condition Room Temperature
Molecular Formula C23 H26 O3
Certifications/Compliance ISO 17034
Synonym Cyclopropanecarboxylic acid, 2,2-dimethyl-3-(2-methyl-1-propen-1-yl)-, (3-phenoxyphenyl)methyl ester, Cyclopropanecarboxylic acid, 2,2-dimethyl-3-(2-methyl-1-propenyl)-, (3-phenoxyphenyl)methyl ester (9CI), Cyclopropanecarboxylic acid, 2,2-dimethyl-3-(2-methylpropenyl)-, m-phenoxybenzyl ester (8CI), Benzyl alcohol, m-phenoxy-, 2,2-dimethyl-3-(2-methylpropenyl)cyclopropanecarboxylate (8CI), 3-Phenoxybenzyl chrysanthemate, Anchimanaito 20S, Duet, Multicide 2154, OMS 1809, OMS 1810, PT 515, Phenothrin, Phenoxythrin, Pibutin, S 2539, S 2539 (pesticide), m-Phenoxybenzyl 2,2-dimethyl-3-(2-methylpropenyl)cyclopropanecarboxylate
InChI Formula InChI=1S/C23H26O3/c1-16(2)13-20-21(23(20,3)4)22(24)25-15-17-9-8-12-19(14-17)26-18-10-6-5-7-11-18/h5-14,20-21H,15H2,1-4H3
Solution Type Neat
SMILES CC(=CC1C(C(=O)OCc2cccc(Oc3ccccc3)c2)C1(C)C)C
IUPAC Name (3-phenoxyphenyl)methyl 2,2-dimethyl-3-(2-methylprop-1-enyl)cyclopropane-1-carboxylate
Industry Type Environmental Testing
Molecular Weight (g/mol) 350.45
Quantity 100 mg
Formula Weight 350.1882 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.