missing translation for 'onlineSavingsMsg'
Learn More

Phenoxyacetic acid, 98+%, Thermo Scientific Chemicals

Product Code. 11945121 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
100 g
500 g
2.5 kg
Packaging:
Glass Bottle
Plastic bottle
Unit Size:
100g
2.5kg
500g
5g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
11945121 2.5 kg Plastic bottle 2.5kg
10002190 5 g Glass Bottle 5g
10687332 100 g Plastic bottle 100g
10307400 500 g Plastic bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 11945121 Supplier Thermo Scientific Acros Supplier No. 169870025
 Restricted Item
Estimated Shipment: 07-12-2026
Ajouter au panier
Ajouter au panier
Request Bulk or Custom Format

missing translation for 'validMainframeMsg'

Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.

CAS: 122-59-8 | C8H8O3 | 152.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Identifiants chimiques

CAS 122-59-8
Molecular Formula C8H8O3
Molecular Weight (g/mol) 152.15
MDL Number MFCD00004296
InChI Key LCPDWSOZIOUXRV-UHFFFAOYSA-N
Synonym phenoxyacetic acid, phenoxyethanoic acid, acetic acid, phenoxy, glycolic acid phenyl ether, phenoxy acetic acid, glycol acid phenyl ether, o-phenylglycolic acid, phenoxyacetate, glycollic acid phenyl ether, acide phenoxyacetique
PubChem CID 19188
ChEBI CHEBI:8075
IUPAC Name 2-phenoxyacetic acid
SMILES OC(=O)COC1=CC=CC=C1

Spécification

Melting Point 98.0°C to 101.0°C
Color White
Boiling Point 285.0°C
Flash Point 165°C
Infrared Spectrum Authentic
Assay Percent Range 98% min. (Acid)
Packaging Plastic bottle
Linear Formula C6H5OCH2CO2H
Quantity 2.5 kg
Beilstein 06,161
Fieser 09,361
Merck Index 15,7366
Solubility Information Solubility in water: soluble. Other solubilities: soluble in glacial acetic acid, soluble in ether, alcohol and carbon disulfide
Formula Weight 152.15
Percent Purity 98+%
Physical Form Granular Powder, Granules, or Prills
Chemical Name or Material Phenoxyacetic acid
Afficher plus Afficher moins
compliance-icons
Identifiant produit
  • Phenoxyacetic acid
Mention d'avertissement
  • Warning
Catégorie de danger
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Mention de danger
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Conseils de prudence
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.