missing translation for 'onlineSavingsMsg'
Learn More

Phenthoate 100 μg/mL in Isooctane, Dr. Ehrenstorfer

Product Code. 30323012
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30323012 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30323012 Supplier Dr. Ehrenstorfer Supplier No. DREXA16050000IO
 Restricted Item
Estimated Shipment: 21-09-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Phenthoate
CAS 2597-03-7
Analyte or Component Names Phenthoate
Shipping Condition Room Temperature
Molecular Formula C12 H17 O4 P S2
Concentration 100 μg/mL
Certifications/Compliance ISO/IEC 17025
Synonym Acetic acid, mercaptophenyl-, ethyl ester O,O-dimethyl phosphorodithioate (6CI), Acetic acid, mercaptophenyl-, ethyl ester, S-ester with O,O-dimethyl phosphorodithioate (7CI), Acetic acid, mercaptophenyl-, ethyl ester, S-ester with O,O-dimethyl phosphorodithioate (8CI), Phosphorodithioic acid, O,O-dimethyl ester, S-ester with ethyl mercaptophenylacetate (8CI), BAY 33051, Bayer 18,510, Bayer 18510, Cidemul, Cidial, Dhanusan 50, Dimephenthoate, ENT 23438, ENT 27386GC, Elsan, Fenthoate, L 561, Montecatini L 561, O,O-Dimethyl S-α-ethoxycarbonylbenzyl phosphorodithioate, Pap, Papthion, Phendal, Phenthoate, S-[α-(ethoxycarbonyl)benzyl] O,O-dimethyl phosphorodithioate, S-α-Ethoxycarbonylbenzyl dimethyl phosphorothiolothionate, Shaluosong, Tagson, Tsidial, Benzeneacetic acid, α-[(dimethoxyphosphinothioyl)thio]-, ethyl ester
InChI Formula InChI=1S/C12H17O4PS2/c1-4-16-12(13)11(10-8-6-5-7-9-10)19-17(18,14-2)15-3/h5-9,11H,4H2,1-3H3
Solution Type Single Solution
SMILES CCOC(=O)C(SP(=S)(OC)OC)c1ccccc1
IUPAC Name ethyl 2-dimethoxyphosphinothioylsulfanyl-2-phenylacetate
Solvent or Matrix Isooctane
Industry Type Environmental Testing
Molecular Weight (g/mol) 320.36
Quantity 1 mL
Formula Weight 320.0306 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.