missing translation for 'onlineSavingsMsg'
Learn More

Phenyl 4-Aminosalicylate, TRC

Product Code. 30417282
Change view
Click to view available options
Quantity:
250 mg
2.5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30417282 250 mg
30421048 2.5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30417282 Supplier Toronto Research Chemicals Supplier No. TRCP309020250MG
 Restricted Item
Estimated Shipment: 04-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 133-11-9
Molecular Formula C13H11NO3
Molecular Weight (g/mol) 229.23
Synonym FR 7, Fenamisal, NSC 40144, PAS-Tebamin, Pheny-PAS-Tebamin, Phenyl 4-Amino-2-hydroxybenzoate, Phenyl 4-Aminosalicylate, Phenyl PAS, Phenyl Aminosalicylate, Phenyl p-Aminosalicylate, Phenyl-PAS-Tebamin, Tebamin, Tebamin-Leo, Tebanyl, p-Aminosalicylic Acid Phenyl Ester, p-Aminosalol, 4-Amino-2-hydroxybenzoic Acid Phenyl Ester, 4-Aminosalicylic Acid Phenyl Ester
IUPAC Name phenyl 4-amino-2-hydroxybenzoate
SMILES Nc1ccc(C(=O)Oc2ccccc2)c(O)c1

Specifications

Quantity 250 mg
InChI Formula InChI=1S/C13H11NO3/c14-9-6-7-11(12(15)8-9)13(16)17-10-4-2-1-3-5-10/h1-8,15H,14H2
Formula Weight 229.0739
Chemical Name or Material Phenyl 4-Aminosalicylate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.