missing translation for 'onlineSavingsMsg'
Learn More

Phenyl 4-Bromo-2-ethylpyridine-1(2H)-carboxylate, TRC

Product Code. 30420151
Change view
Click to view available options
Quantity:
50 mg
500 mg
1 g
Unit Size:
1g
500mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30420151 500 mg 500mg
30420470 50 mg 50mg
30413966 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30420151 Supplier Toronto Research Chemicals Supplier No. TRCP399935500MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Phenyl 4-Bromo-2-ethylpyridine-1(2H)-carboxylate
Analyte or Component Names Phenyl 4-Bromo-2-ethylpyridine-1(2H)-carboxylate
Shipping Condition Room Temperature
Molecular Formula C14 H14 Br N O2
InChI Formula InChI=1S/C14H14BrNO2/c1-2-12-10-11(15)8-9-16(12)14(17)18-13-6-4-3-5-7-13/h3-10,12H,2H2,1H3
Solution Type Neat
SMILES CCC1C=C(Br)C=CN1C(=O)Oc2ccccc2
IUPAC Name phenyl 4-bromo-2-ethyl-2H-pyridine-1-carboxylate
Industry Type Pharmaceutical
Molecular Weight (g/mol) 308.171
Quantity 500 mg
Formula Weight 307.021
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.