missing translation for 'onlineSavingsMsg'
Learn More

Phenyl bromoacetate, 98%, ACROS Organics™

Product Code. 10058329 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 g
50 g
Packaging:
Glass bottle
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10058329 10 g Glass bottle 10g
10483340 50 g Glass bottle 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10058329 Supplier Thermo Scientific Acros Supplier No. 433170100
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 620-72-4
Molecular Formula C8H7BrO2
Molecular Weight (g/mol) 215.046
MDL Number MFCD00192391
InChI Key UEWYUCGVQMZMGY-UHFFFAOYSA-N
Synonym phenyl bromoacetate, acetic acid, bromo-, phenyl ester, bromoacetic acid phenyl ester, bromoacetic acid, phenyl ester, phenylbromessigester, phenyl bromoacetate #, acmc-209mzb, phenyl 2-bromanylethanoate, ksc352o8b
PubChem CID 564919
IUPAC Name phenyl 2-bromoacetate
SMILES C1=CC=C(C=C1)OC(=O)CBr

Specifications

Melting Point 26°C to 33°C
Color White to Yellow
Density 1.508g/mL
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Quantity 10 g
Formula Weight 215.05
Percent Purity ≥97.5%
Physical Form Liquid or Low Melting Solid
Chemical Name or Material Phenyl bromoacetate
Show More Show Less
compliance-icons
Product Identifier
  • Phenyl bromoacetate, 98%
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.