missing translation for 'onlineSavingsMsg'
Learn More

Phenyl bromoacetate, 98%

Product Code. 15416938
Change view
Click to view available options
Quantity:
5 g
50 g
Unit Size:
50g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15416938 50 g 50g
15406938 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15416938 Supplier Thermo Scientific Alfa Aesar Supplier No. H60464.18
 Restricted Item
Estimated Shipment: 31-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 620-72-4 | C8H7BrO2 | 215.046 g/mol

Phenyl bromoacetate may be employed as alkylation reagent in the preparation of 2-(phenoxycarbonyl)methyl triazoles. It may be used in the synthesis of the A-ring of cylindrospermopsin and in the synthesis of 2-{2-[1-(4-hydroxy-6-methyl-2-oxo-2H-pyran-3-yl)ethylidene]hydrazinyl}-1,3-thiazolidin-4-one3, 2-{2-[1-(4-hydroxy-6-methyl-2-oxo-2H-pyran-3-yl)ethylidene]hydrazinyl}-3-methyl-1,3-thiazolidin-4-one3, 3-ethyl-2-{2-[1-(4-hydroxy-6-methyl-2-oxo-2H-pyran-3-yl)ethylidene]hydrazinyl}-1,3-thiazolidin-4-one3 and 2-{2-[1-(4-hydroxy-6-methyl-2-oxo-2H-pyran-3-yl)ethylidene] hydrazinyl}-3-phenyl-1,3-thiazolidin-4-one3.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Phenyl bromoacetate may be employed as alkylation reagent in the preparation of 2-(phenoxycarbonyl)methyl triazoles. It may be used in the synthesis of the A-ring of cylindrospermopsin and in the synthesis of 2-{2-[1-(4-hydroxy-6-methyl-2-oxo-2H-pyran-3-yl)ethylidene]hydrazinyl}-1,3-thiazolidin-4-one3, 2-{2-[1-(4-hydroxy-6-methyl-2-oxo-2H-pyran-3-yl)ethylidene]hydrazinyl}-3-methyl-1,3-thiazolidin-4-one3, 3-ethyl-2-{2-[1-(4-hydroxy-6-methyl-2-oxo-2H-pyran-3-yl)ethylidene]hydrazinyl}-1,3-thiazolidin-4-one3 and 2-{2-[1-(4-hydroxy-6-methyl-2-oxo-2H-pyran-3-yl)ethylidene] hydrazinyl}-3-phenyl-1,3-thiazolidin-4-one3.

Solubility
Soluble in ethanol and ether, insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store under inert gas.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 620-72-4
Molecular Formula C8H7BrO2
Molecular Weight (g/mol) 215.046
MDL Number MFCD00192391
InChI Key UEWYUCGVQMZMGY-UHFFFAOYSA-N
Synonym phenyl bromoacetate, acetic acid, bromo-, phenyl ester, bromoacetic acid phenyl ester, bromoacetic acid, phenyl ester, phenylbromessigester, phenyl bromoacetate #, acmc-209mzb, phenyl 2-bromanylethanoate, ksc352o8b
PubChem CID 564919
IUPAC Name phenyl 2-bromoacetate
SMILES C1=CC=C(C=C1)OC(=O)CBr

Specifications

Melting Point 31°C to 33°C
Density 1.508
Boiling Point 134°C
Flash Point 113°C (235°F)
Linear Formula BrCH2CO2C6H5
Quantity 50 g
Solubility Information Soluble in ethanol and ether,insoluble in water.
Formula Weight 215.04
Percent Purity 98%
Chemical Name or Material Phenyl bromoacetate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.