missing translation for 'onlineSavingsMsg'
Learn More

Phenyl chloroformate, 99%

Product Code. 11444040
Change view
Click to view available options
Quantity:
100 g
250 g
500 g
1000 g
Unit Size:
1000g
100g
250g
500g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11444040 100 g 100g
11450443 250 g 250g
11458680 500 g 500g
11440443 1000 g 1000g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 11444040 Supplier Thermo Scientific Alfa Aesar Supplier No. A12618.22
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1885-14-9 | C7H5ClO2 | 156.57 g/mol

Phenyl chloroformate is used in the synthesis of poly(2-(phenoxycarbonyloxy)ethyl methacrylate) and phenyl-(4-vinylphenyl) carbonate. It acts as a precursor of phenyl mixed anhydrides which are used in peptide coupling reactions. It serves as a dehydrating reagent for the conversion of primary amides to nitriles and an intermediate in the synthesis of pharmaceuticals and carbamates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Phenyl chloroformate is used in the synthesis of poly(2-(phenoxycarbonyloxy)ethyl methacrylate) and phenyl-(4-vinylphenyl) carbonate. It acts as a precursor of phenyl mixed anhydrides which are used in peptide coupling reactions. It serves as a dehydrating reagent for the conversion of primary amides to nitriles and an intermediate in the synthesis of pharmaceuticals and carbamates.

Solubility
Miscible with N,N-dimethylformamide.

Notes
Store in cool place. Moisture sensitive. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with bases, alcohols and amines. Hydrolyses in water.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1885-14-9
Molecular Formula C7H5ClO2
Molecular Weight (g/mol) 156.57
MDL Number MFCD00000637
InChI Key AHWALFGBDFAJAI-UHFFFAOYSA-N
Synonym phenyl chloroformate, chloroformic acid phenyl ester, phenyl chlorocarbonate, carbonochloridic acid, phenyl ester, phenoxycarbonyl chloride, formic acid, chloro-, phenyl ester, fenylester kyseliny chlormravenci, phenylchloroformate, unii-6tnd0d6d3y, 6tnd0d6d3y
PubChem CID 15891
IUPAC Name phenyl carbonochloridate
SMILES ClC(=O)OC1=CC=CC=C1

Specifications

Melting Point -38°C
Density 1.246
Boiling Point 188°C to 189°C
Flash Point 76°C (169°F)
Refractive Index 1.511
Quantity 100 g
UN Number UN2746
Beilstein 606778
Sensitivity Moisture sensitive
Solubility Information Miscible with N,N-dimethylformamide.
Formula Weight 156.57
Percent Purity 99%
Chemical Name or Material Phenyl chloroformate
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Phenyl chloroformate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 2
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Serious eye damage/eye irritation Category 1
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 1 C
  • Specific target organ toxicity Category 3
Hazard Statement
  • H290-May be corrosive to metals.
  • H302-Harmful if swallowed.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • H330-Fatal if inhaled.
  • H335-May cause respiratory irritation.
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contain: Phenyl chloroformate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.