missing translation for 'onlineSavingsMsg'
Learn More

Phenyl chloroformate, 99%

Product Code. 10124560 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 mL
1 L
Packaging:
Glass bottle
Unit Size:
1 L
250mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10124560 250 mL Glass bottle 250mL
11924941 1 L Glass bottle 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.
Product Code. 10124560 Supplier Thermo Scientific Acros Supplier No. 218212500
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.

CAS: 1885-14-9 | C7H5ClO2 | 156.57 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Coupling reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1885-14-9
Molecular Formula C7H5ClO2
Molecular Weight (g/mol) 156.57
MDL Number MFCD00000637
InChI Key AHWALFGBDFAJAI-UHFFFAOYSA-N
Synonym phenyl chloroformate, chloroformic acid phenyl ester, phenyl chlorocarbonate, carbonochloridic acid, phenyl ester, phenoxycarbonyl chloride, formic acid, chloro-, phenyl ester, fenylester kyseliny chlormravenci, phenylchloroformate, unii-6tnd0d6d3y, 6tnd0d6d3y
PubChem CID 15891
IUPAC Name phenyl carbonochloridate
SMILES ClC(=O)OC1=CC=CC=C1

Specifications

Melting Point -28.0°C
Density 1.2440g/mL
Boiling Point 188.0°C to 189.0°C (1013.0 mbar)
Flash Point 80°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula ClCO2C6H5
Refractive Index 1.5100 to 1.5120
Quantity 250 mL
Specific Gravity 1.244
Solubility Information Solubility in water: hydrolysis
Viscosity 1.86 mPa.s (20°C)
Formula Weight 156.57
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Phenyl chloroformate
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Phenyl chloroformate
Signal Word
  • Danger
Hazard Category
  • Skin corrosion/irritation Category 1 B
  • Skin sensitiser Category 1
  • Acute toxicity Category 2
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Substances and mixtures corrosive to metals Category 1
  • Specific target organ toxicity Category 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H314-Causes severe skin burns and eye damage.
  • H317-May cause an allergic skin reaction.
  • H330-Fatal if inhaled.
  • H290-May be corrosive to metals.
  • H412-Harmful to aquatic life with long lasting effects.
  • EUH029-Contact with water liberates toxic gas.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P402+P404-Store in a dry place. Store in a closed container.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.