missing translation for 'onlineSavingsMsg'
Learn More

Phenyl chlorothionoformate, 98+%

Product Code. 11374314
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11374314 1 g 1g
11384314 5 g 5g
11394314 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11374314 Supplier Thermo Scientific Alfa Aesar Supplier No. L00838.03
 Restricted Item
Estimated Shipment: 19-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 1005-56-7 | C7H5ClOS | 172.63 g/mol

Phenyl chlorothionoformate has been used in stereo directed synthesis of optically active, (-)-mintlactone, synthesis of peptide α-thioesters having a variety of C-terminal amino acids, synthesis of scyllo-inositol derivatives, thionocarbonylation of unprotected thymine nucleosides, preparation of phenoxythiocarbonyl esters of protected ribonucleosides. It is also used as a reagent in the thionocarbonylation of unprotected thymine nucleosides. It forms phenoxythiocarbonyl esters of protected ribonucleosides which can be reduced by tributyltin hydride to deoxyribonucleosides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Phenyl chlorothionoformate has been used in stereo directed synthesis of optically active, (-)-mintlactone, synthesis of peptide α-thioesters having a variety of C-terminal amino acids, synthesis of scyllo-inositol derivatives, thionocarbonylation of unprotected thymine nucleosides, preparation of phenoxythiocarbonyl esters of protected ribonucleosides. It is also used as a reagent in the thionocarbonylation of unprotected thymine nucleosides. It forms phenoxythiocarbonyl esters of protected ribonucleosides which can be reduced by tributyltin hydride to deoxyribonucleosides.

Solubility
Soluble in water.

Notes
Moisture, Heat Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents. Stable under recommended storage conditions.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1005-56-7
Molecular Formula C7H5ClOS
Molecular Weight (g/mol) 172.63
MDL Number MFCD00004920
InChI Key KOSYAAIZOGNATQ-UHFFFAOYSA-N
Synonym o-phenyl carbonochloridothioate, phenyl chlorothionoformate, o-phenyl chlorothioformate, phenyl chlorothioformate, phenyl thioxochloroformate, phenyl chlorothionocarbonate, phenoxythiocarbonyl chloride, chlorothioformic acid phenyl ester, o-phenyl chlorothionoformate, o-phenyl chlorothiocarbonate
PubChem CID 70498
SMILES ClC(=S)OC1=CC=CC=C1

Specifications

Density 1.248
Boiling Point 81°C to 83°C (6 mmHg)
Flash Point 81°C (177°F)
Odor Acrid
Refractive Index 1.5804
Quantity 1 g
UN Number UN3265
Beilstein 774830
Sensitivity Moisture sensitive
Solubility Information Soluble in water.
IUPAC Name phenyl chloromethanethioate
Formula Weight 172.63
Percent Purity ≥98%
Chemical Name or Material Phenyl chlorothionoformate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.