missing translation for 'onlineSavingsMsg'
Learn More

Phenyl dichlorophosphate, 99%

Product Code. 10113520 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10113520 25 g Glass bottle 25g
10521554 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10113520 Supplier Thermo Scientific Acros Supplier No. 130480250
 Restricted Item
Estimated Shipment: 05-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 770-12-7 | C6H5Cl2O2P | 210.98 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 770-12-7
Molecular Formula C6H5Cl2O2P
Molecular Weight (g/mol) 210.98
MDL Number MFCD00002067
InChI Key TXFOLHZMICYNRM-UHFFFAOYSA-N
Synonym phenyl dichlorophosphate, phenyl phosphorodichloridate, dichlorophenoxyphosphine oxide, phosphorodichloridic acid, phenyl ester, phenylphosphoric dichloride, phenyl phosphorodichlorodate, phenoxydichlorophosphine oxide, phenyldichlorophosphate, phenoxyphosphoryl dichloride, phosphoroyl dichloride phenyl ester
PubChem CID 13038
IUPAC Name dichlorophosphoryloxybenzene
SMILES C1=CC=C(C=C1)OP(=O)(Cl)Cl

Specifications

Melting Point -1°C
Color Brown to Colorless
Density 1.4120g/mL
Boiling Point 241°C to 243°C
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula C6H5OP(O)Cl2
Refractive Index 1.5210 to 1.523
Quantity 25 g
Beilstein 06, 179
Fieser 01,847; 11,410; 12,384; 13,240; 15,146
Merck Index 15, 7394
Specific Gravity 1.412
Solubility Information Solubility in water: decomposes
Formula Weight 210.98
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Phenyl dichlorophosphate, 99%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.