missing translation for 'onlineSavingsMsg'
Learn More

Phenyl ether, 99%

Product Code. 10183903 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 g
500 g
1 kg
2.5 kg
10 kg
Packaging:
Glass bottle
Plastic drum
Unit Size:
100g
10kg
1kg
2.5kg
500g
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10183903 100 g Glass bottle 100g
10235420 500 g Glass bottle 500g
10346920 1 kg Glass bottle 1kg
10627753 2.5 kg Glass bottle 2.5kg
11924711 10 kg Plastic drum 10kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 10183903 Supplier Thermo Scientific Acros Supplier No. 130601000
 Restricted Item
Estimated Shipment: 19-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 101-84-8 | C12H10O | 170.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 101-84-8
Molecular Formula C12H10O
Molecular Weight (g/mol) 170.21
MDL Number MFCD00003034
InChI Key USIUVYZYUHIAEV-UHFFFAOYSA-N
Synonym diphenyl ether, diphenyl oxide, phenyl ether, oxydibenzene, phenyl oxide, oxybisbenzene, biphenyl oxide, 1,1'-oxydibenzene, benzene, 1,1'-oxybis, oxydiphenyl
PubChem CID 7583
ChEBI CHEBI:39258
IUPAC Name phenoxybenzene
SMILES C1=CC=C(C=C1)OC2=CC=CC=C2

Specifications

Melting Point 26.0°C to 30.0°C
Density 1.0730g/mL
Boiling Point 259.0°C
Flash Point 115°C
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula (C6H5)2O
Refractive Index 1.5795 to 1.5815
Quantity 100 g
Beilstein 06,146
Merck Index 15,3357
Specific Gravity 1.073
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in alcohol,benzene,ether and,glacial acetic acid
Viscosity 3.4909 mPa.s (28°C)
Formula Weight 170.21
Percent Purity 99%
Chemical Name or Material Phenyl ether
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.