missing translation for 'onlineSavingsMsg'
Learn More

Phenyl Ether, TRC

Product Code. 30137006
Change view
Click to view available options
Quantity:
5 g
50 g
100 g
Unit Size:
100g
50g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30137006 50 g 50g
30134831 100 g 100g
30011139 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30137006 Supplier Toronto Research Chemicals Supplier No. TRCP32133050G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 101-84-8
Formula Weight 170.0732
InChI Formula InChI=1S/C12H10O/c1-3-7-11(8-4-1)13-12-9-5-2-6-10-12/h1-10H
IUPAC Name phenoxybenzene
Molecular Formula C12 H10 O
Molecular Weight (g/mol) 170.21
SMILES O(c1ccccc1)c2ccccc2
Synonym Oxydibenzene, Propofol Imp. I (EP), Benzene, 1,1'-oxybis-, Phenyl ether (8CI), 1,1'-Oxybis[benzene], Barrel Therm 330, Benzene, phenoxy-, Biphenyl oxide, Chemcryl JK-EB, Diphenyl ether, Diphenyl oxide, NSC 19311, Oxybisbenzene, Phenoxybenzene, Phenyl oxide
Chemical Name or Material Oxydibenzene

Specifications

Quantity 50 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.