missing translation for 'onlineSavingsMsg'
Learn More

Phenyl p-tolyl sulfone, 98+%

Product Code. p-7043857
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
This item is not returnable. View return policy

Product Code. 11392026

Brand: Thermo Scientific Alfa Aesar L03128.09

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 640-57-3 | C13H12O2S | 232.297 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 640-57-3
Molecular Formula C13H12O2S
Molecular Weight (g/mol) 232.297
MDL Number MFCD00025038
InChI Key YBRXHRWLEFCFEG-UHFFFAOYSA-N
Synonym phenyl p-tolyl sulfone, sulfone, phenyl p-tolyl, 4-methylphenyl phenyl sulfone, benzene, 1-methyl-4-phenylsulfonyl, 1-methyl-4-phenylsulfonyl benzene, 4-methyldiphenylsulfone, p-tolyl phenyl sulfone, p-phenylsulphonyl toluene, 1-benzenesulfonyl-4-methylbenzene, 1-benzenesulfonyl-4-methyl-benzene
PubChem CID 69497
IUPAC Name 1-(benzenesulfonyl)-4-methylbenzene
SMILES CC1=CC=C(C=C1)S(=O)(=O)C2=CC=CC=C2

Specifications

Melting Point 125°C to 129°C
Odor Odorless
Quantity 10 g
Beilstein 1872872
Formula Weight 232.3
Percent Purity ≥98%
Chemical Name or Material Phenyl p-tolyl sulfone

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.