missing translation for 'onlineSavingsMsg'
Learn More

Phenyl phosphate disodium salt hydrate, 98%

Product Code. 11429940
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11429940 100 g 100g
11419940 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11429940 Supplier Thermo Scientific Alfa Aesar Supplier No. 044711.22
 Restricted Item
Estimated Shipment: 05-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Used in phosphatase determinations | CAS: 3279-54-7 | C6H5Na2O4P | 218.06 g/mol

Phenyl phosphate disodium salt is used in phosphatase determinations. It is also used as reagent in chemical research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3279-54-7
Molecular Formula C6H5Na2O4P
Molecular Weight (g/mol) 218.06
MDL Number MFCD00002133
InChI Key TYJOJLOWRIQYQM-UHFFFAOYSA-L
Synonym disodium phenyl phosphate, phenyl phosphate disodium salt, disodium phenylphosphate, phenyldisodium phosphate, phosphoric acid, monophenyl ester, disodium salt, phenyl disodium orthophosphate, phenyl phosphate, phosphoric acid, monophenyl ester, sodium salt 1:2, disodium phenyl phosphate dihydrate, phenyl phosphate disodium salt dihydrate
PubChem CID 76771
IUPAC Name disodium;phenyl phosphate
SMILES [Na+].[Na+].[O-]P([O-])(=O)OC1=CC=CC=C1

Specifications

Color Yellow
Linear Formula C6H5OPO(ONa)2·xH2O
Quantity 100 g
Beilstein 4167154
Sensitivity Hygroscopic
Merck Index 14,3357
Solubility Information Soluble in water (0.1g/mL).
Formula Weight 218.06 (Anhydrous)
Percent Purity 98%
Physical Form Crystalline
Chemical Name or Material Phenyl phosphate disodium salt hydrate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.