missing translation for 'onlineSavingsMsg'
Learn More

Phenyl salicylate, 99%

Product Code. 10552761 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 g
1 kg
2.5 kg
Packaging:
Glass bottle
Plastic Bottle
Unit Size:
1kg
2.5kg
250g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10552761 250 g Glass bottle 250g
10647522 1 kg Glass bottle 1kg
11945091 2.5 kg Plastic Bottle 2.5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10552761 Supplier Thermo Scientific Acros Supplier No. 165082500
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Phenyl salicylate, 99%, C13H10O3, CAS Number-118-55-8, salol, salphenyl, phenyl salicylate, 2-phenoxycarbonylphenol, salicylic acid, phenyl ester, benzoic acid, 2-hydroxy-, phenyl ester, phenyl-2-hydroxybenzoate, musol, phenol salicylate, 2-hydroxybenzoic acid phenyl ester, 250g, 98.5% min. (GC) | CAS: 118-55-8 | C13H10O3 | 214.22 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 118-55-8
Molecular Formula C13H10O3
Molecular Weight (g/mol) 214.22
InChI Key ZQBAKBUEJOMQEX-UHFFFAOYSA-N
Synonym phenyl salicylate, salol, phenol salicylate, salphenyl, musol, 2-phenoxycarbonylphenol, benzoic acid, 2-hydroxy-, phenyl ester, salicylic acid, phenyl ester, 2-hydroxybenzoic acid phenyl ester, phenyl-2-hydroxybenzoate
PubChem CID 8361
ChEBI CHEBI:34918
IUPAC Name phenyl 2-hydroxybenzoate
SMILES C1=CC=C(C=C1)OC(=O)C2=CC=CC=C2O

Specifications

Melting Point 40.0°C to 44.0°C
Color White
Boiling Point 172.0°C to 173.0°C (12.0 mmHg)
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula 2-(HO)C6H4CO2C6H5
Quantity 250 g
Beilstein 10,76
Merck Index 15,742
Solubility Information Solubility in water: <0.1% (25°C). Other solubilities: 1g/1.5mL benzene- 1g/5mL amyl alcohol,53g/100g abs.ethanol-470g/100g ethylacetate (25 c),620g/100g methylethylketone-460g/100g toluene(25 c,soluble in chloroform,acetone,ether and oils,s
Formula Weight 214.22
Percent Purity 99%
Physical Form Fine Crystalline Powder
Chemical Name or Material Phenyl salicylate
Show More Show Less
compliance-icons
Product Identifier
  • Phenyl salicylate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.