missing translation for 'onlineSavingsMsg'
Learn More

Phenyl Salicylate, Extra Pure, SLR, Fisher Chemical

Product Code. 10020290 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
250 g
Packaging:
Plastic powder jar
Unit Size:
250g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10020290 250 g Plastic powder jar 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10020290 Supplier Fisher Chemical Supplier No. P/2840/50
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 118-55-8
Melting Point 40°C
Color White
Boiling Point 172°C
Packaging Plastic powder jar
Molecular Formula C13H10O3
MDL Number 2213
Quantity 250 g
Synonym phenyl salicylate, salol, phenol salicylate, salphenyl, musol, 2-phenoxycarbonylphenol, benzoic acid, 2-hydroxy-, phenyl ester, salicylic acid, phenyl ester, 2-hydroxybenzoic acid phenyl ester, phenyl-2-hydroxybenzoate
InChI Key ZQBAKBUEJOMQEX-UHFFFAOYSA-N
SMILES C1=CC=C(C=C1)OC(=O)C2=CC=CC=C2O
IUPAC Name phenyl 2-hydroxybenzoate
Molecular Weight (g/mol) 214.22
PubChem CID 8361
ChEBI CHEBI:34918
Formula Weight 214.22g/mol
Physical Form Powder
Show More Show Less
compliance-icons
Product Identifier
  • Phenyl salicylate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.