missing translation for 'onlineSavingsMsg'
Learn More

Phenyl trans-beta-styryl sulfone, 96%

Product Code. 11448487
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11448487 5 g 5g
11438487 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11448487 Supplier Thermo Scientific Alfa Aesar Supplier No. A18556.06
 Restricted Item
Estimated Shipment: 03-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 16212-06-9 | C14H12O2S | 244.308 g/mol

Phenyl trans-beta-styryl sulfone is used as a building block for proteomics research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Phenyl trans-beta-styryl sulfone is used as a building block for proteomics research.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16212-06-9
Molecular Formula C14H12O2S
Molecular Weight (g/mol) 244.308
MDL Number MFCD00159177
InChI Key DNMCCXFLTURVLK-VAWYXSNFSA-N
Synonym phenyl trans-styryl sulfone, phenyl styryl sulfone, sulfone, phenyl styryl, phenyl trans-styryl sulphone, phenyl 2-phenylvinyl sulfone, benzene, 2-phenylethenyl sulfonyl, e-2-benzenesulfonyl ethenyl benzene, 1e-2-phenylvinyl sulfonyl benzene, styrylsulfonylbenzene, e-2-phenylethenyl sulphonylbenzene
PubChem CID 736143
IUPAC Name [(E)-2-(benzenesulfonyl)ethenyl]benzene
SMILES C1=CC=C(C=C1)C=CS(=O)(=O)C2=CC=CC=C2

Specifications

Melting Point 74°C to 76°C
Quantity 5 g
Formula Weight 244.32
Percent Purity 98%
Chemical Name or Material Phenyl trans-beta-styryl sulfone

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.