missing translation for 'onlineSavingsMsg'
Learn More

Phenyl trifluoroacetate, 98%, Thermo Scientific™

Product Code. 10185370 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
:
25mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
10185370 25mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 10185370 Supplier Thermo Scientific Acros Supplier No. 309450250
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 500-73-2
Color Colorless
Density 1.2700g/mL
Boiling Point 146.0°C to 147.0°C
Flash Point 52°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass Bottle
Molecular Formula C8H5F3O2
Linear Formula CF3CO2C6H5
Refractive Index 1.4185 to 1.4225
MDL Number MFCD00039232
Quantity 25 mL
Specific Gravity 1.27
Synonym phenyl trifluoroacetate, acetic acid, trifluoro-, phenyl ester, acetic acid, 2,2,2-trifluoro-, phenyl ester, trifluoroacetic acid phenyl ester, phenyltrifluoroacetate, acmc-209kjz, dvcmyaiusosiqp-uhfffaoysa
InChI Key DVCMYAIUSOSIQP-UHFFFAOYSA-N
SMILES C1=CC=C(C=C1)OC(=O)C(F)(F)F
IUPAC Name phenyl 2,2,2-trifluoroacetate
Molecular Weight (g/mol) 190.12
PubChem CID 68143
Formula Weight 190.12
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Phenyl trifluoroacetate, 98%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.