missing translation for 'onlineSavingsMsg'
Learn More

Phenyl trifluoromethyl sulfide, 98%

Product Code. 11338235
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11338235 1 g 1g
11348235 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11338235 Supplier Thermo Scientific Alfa Aesar Supplier No. L10114.03
 Restricted Item
Estimated Shipment: 07-10-2026
Add to basket
This item is not returnable. View return policy

CAS: 456-56-4 | C7H5F3S | 178.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 456-56-4
Molecular Formula C7H5F3S
Molecular Weight (g/mol) 178.17
MDL Number MFCD00040839
InChI Key YQQKTCBMKQQOSM-UHFFFAOYSA-N
Synonym phenyl trifluoromethyl sulfide, trifluoromethylthiobenzene, trifluoromethylthio benzene, trifluoromethyl thio benzene, benzene, trifluoromethyl thio, trifluoromethyl phenyl sulfide, phenyl trifluoromethyl sulphide, pubchem10400, phenyl trifluoromethylsulfide
PubChem CID 68011
IUPAC Name trifluoromethylsulfanylbenzene
SMILES FC(F)(F)SC1=CC=CC=C1

Specifications

Density 1.249
Boiling Point 140°C to 142°C
Flash Point 37°C (98°F)
Odor Unpleasant
Refractive Index 1.4655
Quantity 1 g
UN Number UN1993
Beilstein 2043134
Formula Weight 178.18
Percent Purity 98%
Chemical Name or Material Phenyl trifluoromethyl sulfide
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.