Learn More
Phenylalanine Hydroxylase Antibody (S06-3H4), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand:  Novus Biologicals NBP3-19516-100ul
 View more versions of this product
Product Code. 18681194
  • 645.00 € / 100µL
Description
Phenylalanine Hydroxylase Monoclonal antibody specifically detects Phenylalanine Hydroxylase in Human, Mouse, Rat samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Phenylalanine Hydroxylase
Monoclonal
0.3 mg/mL
50mM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA
Rabbit
Affinity purified
RUO
5053
Store at 4°C short term. Store at -20°C long term. Avoid freeze-thaw cycles.
IgG
Western Blot
S06-3H4
Unconjugated
EC 1.14.16, EC 1.14.16.1, PH, phe-4-monooxygenase, phenylalanine 4-monooxygenase, phenylalanine hydroxylase, phenylalanine-4-hydroxylase, PKU, PKU1
A synthetic peptide of human Phenylalanine Hydroxylase (Uniprot # P00439)
100 μg
Primary
Human, Mouse, Rat
Purified