missing translation for 'onlineSavingsMsg'
Learn More

Phenylethyl isothiocyanate, 96%

Product Code. 10541691 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10541691 25 g Glass bottle 25g
10053220 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10541691 Supplier Thermo Scientific Acros Supplier No. 265160250
 Restricted Item
Estimated Shipment: 30-09-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 2257-09-2 | C9H9NS | 163.24 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2257-09-2
Molecular Formula C9H9NS
Molecular Weight (g/mol) 163.24
MDL Number MFCD00004821
InChI Key IZJDOKYDEWTZSO-UHFFFAOYSA-N
ChEBI CHEBI:351346

Specifications

Color Colorless to Yellow
Density 1.1000g/mL
Boiling Point 113.0°C (1.0 mmHg)
Flash Point 100°C
Infrared Spectrum Authentic
Assay Percent Range 96%
Packaging Glass bottle
Linear Formula C6H5CH2CH2NCS
Refractive Index 1.5860 to 1.5910
Quantity 25 g
Beilstein 12, 1100
Merck Index 15, 7340
Specific Gravity 1.1
Synonym 2-phenylethyl isothiocyanate, phenethyl isothiocyanate, phenylethyl isothiocyanate, 2-isothiocyanatoethyl benzene, phenethyl mustard oil, phenylaethylsenfoel, benzene, 2-isothiocyanatoethyl, peitc, phenylethyl mustard oil, 2-phenylethylisothiocyanate
Solubility Information Solubility in water: insoluble
SMILES C1=CC=C(C=C1)CCN=C=S
IUPAC Name 2-isothiocyanatoethylbenzene
PubChem CID 16741
Formula Weight 163.24
Percent Purity 96%
Physical Form Liquid
Chemical Name or Material Phenylethyl isothiocyanate, 96%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Phenylethyl isothiocyanate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Skin sensitiser Category 1
  • Specific target organ toxicity Category 3
Hazard Statement
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.