missing translation for 'onlineSavingsMsg'
Learn More

Phenylhydrazine, Extra Pure, SLR, Fisher Chemical

Product Code. 10171720 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
250 mL
Packaging:
Plastic coated glass bottle
Unit Size:
250mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10171720 250 mL Plastic coated glass bottle 250mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10171720 Supplier Fisher Chemical Supplier No. P/2560/PB07
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 100-63-0
Melting Point 19°C
Color Amber
Boiling Point 238°C
Packaging Plastic coated glass bottle
Molecular Formula C6H8N2
MDL Number MFCD00007573
Quantity 250 mL
Synonym hydrazinobenzene, hydrazine, phenyl, phenylhydrazin, monophenylhydrazine, phenyl hydrazine, 1-phenylhydrazine, hydrazine-benzene, fenilidrazina, fenylhydrazine, fenylhydrazine dutch
InChI Key HKOOXMFOFWEVGF-UHFFFAOYSA-N
SMILES NNC1=CC=CC=C1
IUPAC Name phenylhydrazine
Molecular Weight (g/mol) 108.14
PubChem CID 7516
ChEBI CHEBI:27924
Formula Weight 108.14g/mol
Physical Form Liquid
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Phenylhydrazine
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • ACUTE AQUATIC Acute 1
  • Carcinogenicity Category 1B
  • Serious eye damage/eye irritation Category 2
  • Germ cell mutagenicity Category 2
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 1
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H372-Causes damage to organs.
  • H400-Very toxic to aquatic life.
  • H350-May cause cancer.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H341-Suspected of causing genetic defects.
  • H311-Toxic in contact with skin.
  • H331-Toxic if inhaled.
  • H301-Toxic if swallowed.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P311-Call a POISON CENTER/doctor/ .
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.