missing translation for 'onlineSavingsMsg'
Learn More

Phenylhydrazine hydrochloride, 99%, Thermo Scientific Chemicals

Product Code. 11417606
Click to view available options
Quantity:
500 g
Unit Size:
500g
This item is not returnable. View return policy

Product Code. 11417606

Brand: Thermo Scientific Alfa Aesar A14645.36

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Phenylhydrazine hydrochloride is used as a selective mannosidase inhibitor. It is used in the study of tyrosine phosphorylation of janus protein tyrosine kinase in the EPO-responsive normal erythroblastoid cells of anemic mice. It is used in steroid assays and as an N-protecting reagent. Its free base, phenylhydrazine is used to prepare indoles, which find application as intermediates in the synthesis of various dyes and pharmaceuticals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Phenylhydrazine hydrochloride is used as a selective mannosidase inhibitor. It is used in the study of tyrosine phosphorylation of janus protein tyrosine kinase in the EPO-responsive normal erythroblastoid cells of anemic mice. It i
TRUSTED_SUSTAINABILITY

Specifications

CAS 59-88-1
Melting Point ∼252°C (decomposition)
Molecular Formula C6H9ClN2
MDL Number MFCD00012924
Quantity 500 g
UN Number UN2811
Beilstein 3594958
Merck Index 14,7293
Synonym phenylhydrazine hydrochloride, phenylhydrazinium chloride, phenylhydrazine-hcl, phenylhydrazine monohydrochloride, phenylhydrazine hcl, hydrazine, phenyl-, monohydrochloride, unii-h7qfk49svd, phenylhydrazin hydrochlorid, phenylhydrazine.hcl, ccris 5707
Solubility Information Soluble aq. soln. and alcohol.
InChI Key JOVOSQBPPZZESK-UHFFFAOYSA-N
SMILES [H+].[Cl-].NNC1=CC=CC=C1
IUPAC Name phenylhydrazine;hydrochloride
Molecular Weight (g/mol) 144.60
PubChem CID 60962
Formula Weight 144.61
Percent Purity 99%
Chemical Name or Material Phenylhydrazine hydrochloride
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.