missing translation for 'onlineSavingsMsg'
Learn More

Phenylpyruvic acid, sodium salt monohydrate, 98%

Product Code. 10082450 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10082450 5 g Glass bottle 5g
10503941 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10082450 Supplier Thermo Scientific Acros Supplier No. 189730050
 Restricted Item
Estimated Shipment: 04-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 122049-54-1 | C9H7O3 | 163.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 122049-54-1
Molecular Formula C9H7O3
Molecular Weight (g/mol) 163.15
MDL Number MFCD00149109
InChI Key BTNMPGBKDVTSJY-UHFFFAOYSA-M
Synonym sodium phenylpyruvate monohydrate, sodium 2-oxo-3-phenylpropanoate hydrate, phenylpyruvic acid sodium salt monohydrate, phenylpyruvic acid, sodium salt, sodium hydrate ?-phenylpyruvate, l-phenyl pyruvic acid sodium salt, beta-phenylpyruvic acid sodium salt, acmc-2099lh, phenylpyruvic acid sodium hydrate, beta-phenylpyruvic acid, sodium salt
PubChem CID 23666336
IUPAC Name sodium;2-oxo-3-phenylpropanoate;hydrate
SMILES [O-]C(=O)C(=O)CC1=CC=CC=C1

Specifications

Color White
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula C6H5CH2COCO2Na·H2O
Quantity 5 g
Beilstein 10, I, 325
Solubility Information Solubility in water: soluble in water
Formula Weight 204.16
Percent Purity 98%
Physical Form Fine Powder
Chemical Name or Material Phenylpyruvic acid, sodium salt monohydrate, 98%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.