missing translation for 'onlineSavingsMsg'
Learn More

(Phenylsulfonyl)acetic acid, 97%

Product Code. 11378085
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11378085 1 g 1g
11388085 5 g 5g
11398085 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11378085 Supplier Thermo Scientific Alfa Aesar Supplier No. L08299.03
 Restricted Item
Estimated Shipment: 08-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 3959-23-7 | C8H8O4S | 200.208 g/mol

(Phenylsulfonyl)acetic acid is used as pharmaceutical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(Phenylsulfonyl)acetic acid is used as pharmaceutical intermediates.

Notes
Incompatible materials are Strong oxidizing agents. Store in cool place. Keep container tightly closed in a dry and well-ventilated place
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3959-23-7
Molecular Formula C8H8O4S
Molecular Weight (g/mol) 200.208
MDL Number MFCD00025042
InChI Key YTEFAALYDTWTLB-UHFFFAOYSA-N
Synonym phenylsulfonyl acetic acid, 2-phenylsulfonyl acetic acid, 2-benzenesulfonyl acetic acid, phenylsulfonylacetic acid, acetic acid, phenylsulfonyl, benzenesulfonyl-acetic acid, benzenesulfonyl acetic acid, 2-phenylsulphonyl acetic acid, benzenesulfonylacetic acid, phenyl sulfonylacetic acid
PubChem CID 59543
IUPAC Name 2-(benzenesulfonyl)acetic acid
SMILES C1=CC=C(C=C1)S(=O)(=O)CC(=O)O

Specifications

Melting Point 109°C to 114°C
Quantity 1 g
Beilstein 2504376
Formula Weight 200.22
Percent Purity 97%
Chemical Name or Material (Phenylsulfonyl)acetic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.