missing translation for 'onlineSavingsMsg'
Learn More

(Phenylthio)acetic acid, 97%

Product Code. 11455946
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11455946 100 g 100g
11445946 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11455946 Supplier Thermo Scientific Alfa Aesar Supplier No. B22280.22
 Restricted Item
Estimated Shipment: 30-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 103-04-8 | C8H7O2S | 167.20 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 103-04-8
Molecular Formula C8H7O2S
Molecular Weight (g/mol) 167.20
MDL Number MFCD00004355
InChI Key MOTOSAGBNXXRRE-UHFFFAOYSA-M
Synonym phenylthio acetic acid, thiophenoxyacetic acid, 2-phenylthio acetic acid, phenylthioglycolic acid, phenylmercaptoacetic acid, 2-phenylsulfanyl acetic acid, phenylsulfanyl acetic acid, acetic acid, phenylthio, phenylmercapto acetic acid, carboxymethyl phenyl sulfide
PubChem CID 59541
IUPAC Name 2-phenylsulfanylacetic acid
SMILES [O-]C(=O)CSC1=CC=CC=C1

Specifica

Melting Point 61°C to 66°C
Odor Stench
Quantity 100 g
Beilstein 2045072
Formula Weight 168.22
Percent Purity 97%
Chemical Name or Material (Phenylthio)acetic acid

RUO – Research Use Only

Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.