missing translation for 'onlineSavingsMsg'
Learn More

Phenylthioacetonitrile, 98%, Thermo Scientific™

Product Code. 10571472 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10571472 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10571472 Supplier Thermo Scientific Acros Supplier No. 355330010

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5219-61-4
Molecular Formula C8H7NS
Molecular Weight (g/mol) 149.21
MDL Number MFCD00019831
InChI Key QMJWPWCKRKHUNY-UHFFFAOYSA-N
Synonym phenylthio acetonitrile, phenylthioacetonitrile, 2-phenylthio acetonitrile, acetonitrile, phenylthio, phenylmercaptoacetonitrile, cyanomethyl phenyl sulfide, 2-phenylsulfanyl acetonitrile, 2-phenylthioethanenitrile, acmc-209ky7, phenylmercapto acetonitrile
PubChem CID 78891
IUPAC Name 2-(phenylsulfanyl)acetonitrile
SMILES N#CCSC1=CC=CC=C1

Specifications

Color Colorless to Yellow
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Refractive Index 1.5790 to 1.5810 (20°C, 589nm)
Quantity 1 g
Formula Weight 149.22
Percent Purity ≥97.5%
Physical Form Liquid
Chemical Name or Material Phenylthioacetonitrile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.