missing translation for 'onlineSavingsMsg'
Learn More

Phenyltrimethylammonium tribromide, 97%

Product Code. 11458226
Change view
Click to view available options
Quantity:
50 g
250 g
1000 g
Unit Size:
1000g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11458226 250 g 250g
11448226 50 g 50g
11438226 1000 g 1000g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11458226 Supplier Thermo Scientific Alfa Aesar Supplier No. A15326.30
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 4207-56-1 | C9H14I3N | 516.931 g/mol

Phenyltrimethylammonium tribromide is used as brominating reagent for 1,2-addition of bromine to the double bond of α,β-unsaturated compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Phenyltrimethylammonium tribromide is used as brominating reagent for 1,2-addition of bromine to the double bond of α,β-unsaturated compounds.

Solubility
Insoluble in water.

Notes
Hygroscopic. Incompatible with oxidizing agents, reducing agents and bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4207-56-1
Molecular Formula C9H14I3N
Molecular Weight (g/mol) 516.931
MDL Number MFCD00011789
InChI Key RHXJPCQWYPXLEH-UHFFFAOYSA-N
Synonym mono n,n,n-trimethylbenzenaminium tribromide
PubChem CID 21525000
IUPAC Name trimethyl(phenyl)azanium;triiodide
SMILES C[N+](C)(C)C1=CC=CC=C1.I[I-]I

Specifications

Melting Point 112°C to 117°C
Odor Irritating
Quantity 250 g
UN Number UN3261
Beilstein 3919459
Sensitivity Hygroscopic
Solubility Information Insoluble in water.
Formula Weight 375.93
Percent Purity 97%
Chemical Name or Material Phenyltrimethylammonium tribromide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.