accessibility menu, dialog, popup

UserName

CAS RN 4207-56-1

IUPAC Name: N,N,N-trimethylanilinium; tribroman-2-uide
Synonyms: tetrabutylammonium tribromide; tribroman-2-uide
Molecular Weight (g/mol): 375.93
Molecular Formula: C9H14Br3N
InChi Key: PRXNKYBFWAWBNZ-UHFFFAOYSA-N
SMILES: Br[Br-]Br.C[N+](C)(C)C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (2)
  • (3)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)