missing translation for 'onlineSavingsMsg'
Learn More

Phenylurea, 97%

Product Code. 11304554
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11304554 100 g 100g
11314554 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11304554 Supplier Thermo Scientific Alfa Aesar Supplier No. L01306.22
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 64-10-8 | C7H8N2O | 136.154 g/mol

Phenyl urea is used in organic synthesis. It acts as an efficient ligand for palladium-catalyzed Heck and Suzuki reactions of aryl bromides and iodides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Phenyl urea is used in organic synthesis. It acts as an efficient ligand for palladium-catalyzed Heck and Suzuki reactions of aryl bromides and iodides.

Solubility
Soluble in water.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 64-10-8
Molecular Formula C7H8N2O
Molecular Weight (g/mol) 136.154
MDL Number MFCD00007944
InChI Key LUBJCRLGQSPQNN-UHFFFAOYSA-N
Synonym 1-phenylurea, n-phenylurea, urea, phenyl, phenylcarbamide, monophenylurea, stabilizer vh, stabilisator vh, phenyl-urea, urea, n-phenyl, phenyl urea
PubChem CID 6145
IUPAC Name phenylurea
SMILES C1=CC=C(C=C1)NC(=O)N

Specifications

Melting Point 144°C to 148°C
Density 1.302
Boiling Point 238°C
Linear Formula C6H5NHCONH2
Quantity 100 g
Beilstein 1934615
Merck Index 14,7319
Solubility Information Soluble in water.
Formula Weight 136.15
Percent Purity 97%
Chemical Name or Material Phenylurea
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.