missing translation for 'onlineSavingsMsg'
Learn More

Phenytoin, MedChemExpress

Product Code. 16462179
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16462179 1 g 1g
16440938 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16462179 Supplier Medchem Express Supplier No. HYB0448
 Restricted Item
Estimated Shipment: 08-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Phenytoin (5,5-Diphenylhydantoin) is a potent Voltage-gated Na+ channels (VGSCs) blocker. Phenytoin has antiepileptic activity and reduces breast tumour growth and metastasis in mice.

Phenytoin (5,5-Diphenylhydantoin) is a potent Voltage-gated Na+ channels (VGSCs) blocker. Phenytoin has antiepileptic activity and reduces breast tumour growth and metastasis in mice.

Specifications

Chemical Name or Material Phenytoin
Physical Form Solid
CAS 57-41-0
Color White
Quantity 1 g
Purity Grade Notes Research
Molecular Formula C15H12N2O2
For Use With (Application) Neuroscience-Neuromodulation
Synonym 5,5-Diphenylhydantoin
Solubility Information DMSO : 50 mg/mL (198.20 mM; Need ultrasonic)
SMILES O=C1NC(C(C2=CC=CC=C2)(C3=CC=CC=C3)N1)=O
Molecular Weight (g/mol) 252.27
Formula Weight 252.27
Percent Purity 99.71%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.