missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MAGEB4 Antibody (OTI2H5), Novus Biologicals Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-44721
This item is not returnable.
View return policy
Description
MAGEB4 Monoclonal antibody specifically detects MAGEB4 in Human samples. It is validated for Western Blot
Specifications
| MAGEB4 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized powder ( 1X PBS, pH 7.3,) | |
| Mouse | |
| >95% | |
| RUO | |
| 4115 | |
| Human | |
| Antibody | |
| IgG1 |
| Western Blot | |
| OTI2H5 | |
| Western Blot 1:2000 | |
| cancer/testis antigen family 3, member 6, CT3.6, MAGE-B4 antigen, melanoma antigen family B, 4, melanoma-associated antigen B4, MGC33144 | |
| Human recombinant protein fragment corresponding to amino acids 1-193 of human MAGEB4(NP_002358) produced in E.coli. | |
| 100 μg | |
| Primary | |
| For reconstitution, we recommend adding 100uL distilled water to a final antibody concentration of about 1 mg/mL. To use this carrier-free antibody for conjugation experiment, we strongly recommend performing another round of desalting process. | |
| Store at -20 degrees C. Avoid freeze/thaw cycles. | |
| Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction