missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PHF22 Recombinant Rabbit Monoclonal Antibody (2D3T4)

Rabbit Recombinant Monoclonal Antibody
176.00 € - 436.00 €
Specifications
| Antigen | PHF22 |
|---|---|
| Clone | 2D3T4 |
| Concentration | 0.58 mg/mL |
| Content And Storage | -20°C, Avoid Freeze/Thaw Cycles |
| Applications | ELISA, Western Blot |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30284061
|
Invitrogen™
MA557955 |
20 μL |
176.00 €
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
30282830
![]() |
Invitrogen™
MA557956 |
100 μL |
436.00 €
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Immunogen sequence: MAATVNLELD PIFLKALGFL HSKSKDSAEK LKALLDESLA RGIDSSYRPS QKDVEPPKIS STKNISIKQE PKISSSLPSG NNNGKVLTTE KVKKEAEKRP ADKMKSDITE GVDIPKKPRL EKPETQSSPI TVQSSKDLPM ADLSSFEETS ADDFAMEMGL ACVVCRQMMV ASGNQLVECQ ECHNLYHRDC HKPQVTDKEA NDPRLVWYCA RCTRQMKRMA QKTQKPPQKP.
INTS12 is a subunit of the Integrator complex, which associates with the C-terminal domain of RNA polymerase II large subunit and mediates 3-prime end processing of small nuclear RNAs U1 and U2.Specifications
| PHF22 | |
| 0.58 mg/mL | |
| ELISA, Western Blot | |
| Unconjugated | |
| Rabbit | |
| RUO | |
| PBS with 0.05% BSA, 50% glycerol and 0.05% ProClin 300; pH 7.3 | |
| Q9D168 | |
| 71793 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-230 of human INTS12 (NP_065128.2). | |
| Antibody |
| 2D3T4 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Recombinant Monoclonal | |
| Liquid | |
| IgG | |
| Mouse | |
| Ints12 | |
| 1110020M19Rik; 2810027J24Rik; 4930529N21Rik; A230056J18Rik; AI666778; hypothetical nuclear factor SBBI22; hypothetical protein LOC415251; int12; Integrator complex subunit 12; INTS12; PHD finger protein 22; PHF22; SBBI22; Unknown (protein for MGC:134302); wu:fi17f05; zgc:86602 | |
| Ints12 | |
| Primary | |
| Protein A |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title