missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PHF8 Recombinant Rabbit Monoclonal Antibody (24GB945)
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA557154
This item is not returnable.
View return policy
Description
PHF8 Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow
PHD finger protein 8 is a jumonji domain containing protein. Like other members of the jumonji family, PHF8 may therefore play a role in histone demethylation. Mutations in PHF8 lead to Siderius type X-linked mental retardation (MRXSSD), a mild to borderline type of mental retardation.
Specifications
| PHF8 | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q80TJ7, Q9UPP1 | |
| PHF8 | |
| A synthesized peptide derived from human PHF8 (aa 800-860). | |
| 20 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 24GB945 | |
| Unconjugated | |
| PHF8 | |
| 9830141C09Rik; histone lysine demethylase PHF8; JHDM1F; jumonji C domain-containing histone demethylase 1F; Kiaa1111; mKIAA1111; MRXSSD; PHD finger protein 8; PHF8; RGD1561065; ZNF422 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 23133, 317425, 320595 | |
| -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction