Learn More
PHGDH Antibody (CL0555) [Janelia Fluor™ 669], Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP2-52939JF669
Product Code. 30299551
  • 737.00 € / 0.10mL
Description
PHGDH Monoclonal antibody specifically detects PHGDH in Human samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin),Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY
Specifications
Specifications
PHGDH
Monoclonal
Janelia Fluor 669
3,3-phosphoglycerate dehydrogenase, 3PGDH, 3-PGDH, D-3-phosphoglycerate dehydrogenase, EC 1.1.1, EC 1.1.1.95, MGC3017, PDG, PGAD, PGD, PGDH, phosphoglycerate dehydrogenase, SERA
This antibody was developed using a recombinant protein derived from O43175, with the exact immunogen sequence remaining proprietary.
0.1 mL
Stem Cell Markers
26227
Store at 4⪚C in the dark.
IgG1
Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry/Immunofluorescence
CL0555
50mM Sodium Borate
Mouse
Protein A purified
RUO
Primary
Human
Purified