missing translation for 'onlineSavingsMsg'
Learn More

Melford Phleomycin

Product Code. p-200087134
Click to view available options
Quantity:
5 mg
25 mg
100 mg
250 mg
Unit Size:
100mg
250mg
25mg
5mg
This item is not returnable. View return policy

Product Code. 30018651

Brand: Melford P202000.25

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Phleomycin is an antibiotic agent. It is known for its ability to cleave DNA, making it useful for studies involving DNA damage and repair mechanisms.

Phleomycin, reconstituted, is often employed to induce controlled DNA damage in cells by causing single- and double-strand breaks in the DNA structure. Typically used to investigate cellular responses to DNA damage, as well as to explore the mechanisms involved in DNA repair.

It is a valuable tool for understanding various aspects of molecular biology, including gene regulation and genome integrity.

Specifications

Chemical Name or Material Phleomycin
CAS 11006-33-0
Quantity 250 mg
Molecular Formula C55H85O21N20S2Cu-HCl
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.