missing translation for 'onlineSavingsMsg'
Learn More

Melford Phleomycin

Product Code. 30018651
Change view
Click to view available options
Quantity:
5 mg
25 mg
100 mg
250 mg
Unit Size:
100mg
250mg
25mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30018651 250 mg 250mg
30018648 5 mg 5mg
30018649 25 mg 25mg
30018650 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 30018651 Supplier Melford Supplier No. P202000.25

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Phleomycin is an antibiotic agent. It is known for its ability to cleave DNA, making it useful for studies involving DNA damage and repair mechanisms.

Phleomycin, reconstituted, is often employed to induce controlled DNA damage in cells by causing single- and double-strand breaks in the DNA structure. Typically used to investigate cellular responses to DNA damage, as well as to explore the mechanisms involved in DNA repair.

It is a valuable tool for understanding various aspects of molecular biology, including gene regulation and genome integrity.

Specifications

Chemical Name or Material Phleomycin
CAS 11006-33-0
Quantity 250 mg
Molecular Formula C55H85O21N20S2Cu-HCl
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.