missing translation for 'onlineSavingsMsg'
Learn More

Phloretin, MedChemExpress

Product Code. p-200099374
Encompass_Preferred
Click to view available options
Quantity:
250 mg
500 mg
Unit Size:
250mg
500mg
This item is not returnable. View return policy

Product Code. 16479895

Brand: Medchem Express HYN0142

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Phloretin (NSC 407292; RJC 02792) is a flavonoid extracted from Prunus mandshurica, has anti-inflammatory activities.

Phloridzin is a specific, competitive and orally active inhibitor of sodium/glucose cotransporters in the intestine (SGLT1) and kidney (SGLT2). Phloretin inhibits Yeast-made GLUT1 as well as Human erythrocyte GLUT1 with IC50values of 49 μM and 61 μM, respectively.Phloretin has the potential for the treatment of rheumatoid arthritis (RA) and allergic airway inflammation.

Specifications

Chemical Name or Material Phloretin
CAS 60-82-2
Color Pink
Purity Grade Notes Research
Molecular Formula C15H14O5
Quantity 250 mg
Synonym NSC 407292 RJC 02792
Solubility Information DMSO : 50 mg/mL (182.30 mM; Need ultrasonic) ∣H2O : < 0.1 mg/mL (insoluble)
SMILES O=C(C1=C(O)C=C(O)C=C1O)CCC2=CC=C(O)C=C2
Molecular Weight (g/mol) 274.27
Formula Weight 274.27
Percent Purity 99.78%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.