missing translation for 'onlineSavingsMsg'
Learn More

Phloroglucide, 95%

Product Code. 11445337
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11445337 25 g 25g
11435337 5 g 5g
11455337 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11445337 Supplier Thermo Scientific Alfa Aesar Supplier No. B22693.14
 Restricted Item
Estimated Shipment: 01-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 491-45-2 | C12H10O5 | 234.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 491-45-2
Molecular Formula C12H10O5
Molecular Weight (g/mol) 234.21
MDL Number MFCD01543374
InChI Key KICYRZIVKKYRFS-UHFFFAOYSA-N
Synonym phloroglucide, 2,3',4,5',6-biphenylpentol, 2-3,5-dihydroxyphenyl benzene-1,3,5-triol, 1,1'-biphenyl-2,3',4,5',6-pentol, phloroglucide hydrate, 1,1'-biphenyl-2,3',4,5',6-pentaol, phloroglucid, d0o8yz, 2,4,5',6-biphenylpentol
PubChem CID 248349
SMILES OC1=CC(O)=C(C(O)=C1)C1=CC(O)=CC(O)=C1

Specifications

Melting Point ∼296°C (decomposition)
Quantity 25 g
Beilstein 3324960
Sensitivity Light sensitive
IUPAC Name 2-(3,5-dihydroxyphenyl)benzene-1,3,5-triol
Formula Weight 234.21
Percent Purity 95%
Chemical Name or Material Phloroglucide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.