missing translation for 'onlineSavingsMsg'
Learn More

Phloroglucide hydrate, 95%

Product Code. 10328420 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
Packaging:
Glass bottle
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10328420 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10328420 Supplier Thermo Scientific Acros Supplier No. 241770250
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 491-45-2 | C12H10O5 | 234.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 491-45-2
Melting Point 300°C
Color Yellow to Brown
Infrared Spectrum Authentic
Assay Percent Range 94% min. (HPLC)
Packaging Glass bottle
Molecular Formula C12H10O5
MDL Number MFCD01543374
Quantity 25 g
Synonym phloroglucide, 2,3',4,5',6-biphenylpentol, 2-3,5-dihydroxyphenyl benzene-1,3,5-triol, 1,1'-biphenyl-2,3',4,5',6-pentol, phloroglucide hydrate, 1,1'-biphenyl-2,3',4,5',6-pentaol, phloroglucid, d0o8yz, 2,4,5',6-biphenylpentol
InChI Key KICYRZIVKKYRFS-UHFFFAOYSA-N
SMILES OC1=CC(O)=C(C(O)=C1)C1=CC(O)=CC(O)=C1
IUPAC Name 2-(3,5-dihydroxyphenyl)benzene-1,3,5-triol
Molecular Weight (g/mol) 234.21
PubChem CID 248349
Formula Weight 234.21
Percent Purity 95%
Physical Form Powder
Chemical Name or Material Phloroglucide hydrate, 95%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.