missing translation for 'onlineSavingsMsg'
Learn More

Phloroglucide Hydrate, TRC

Product Code. 30136189
Change view
Click to view available options
Quantity:
1 g
10 g
25 g
Unit Size:
10g
1g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30136189 10 g 10g
30136541 25 g 25g
17982215 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30136189 Supplier Toronto Research Chemicals Supplier No. TRCP33995010G
 Restricted Item
Estimated Shipment: 20-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

Molecular Formula C12 H10 O5 . x(H2O)
Molecular Weight (g/mol) 234.2
Synonym [1,1'-Biphenyl]-2,3',4,5',6-pentol Hydrate, 2,4,6,3',5'-Biphenylpentol Hydrate, NSC 65069 Hydrate;
IUPAC Name 2-(3,5-dihydroxyphenyl)benzene-1,3,5-triol;hydrate
SMILES OC1=CC(O)=CC(O)=C1C2=CC(O)=CC(O)=C2.O

Specifications

Quantity 10 g
InChI Formula InChI=1S/C12H10O5.H2O/c13-7-1-6(2-8(14)3-7)12-10(16)4-9(15)5-11(12)17;/h1-5,13-17H;1H2
Chemical Name or Material Phloroglucide Hydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.