missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PHLPP2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-13757
This item is not returnable.
View return policy
Description
PHLPP2 Polyclonal antibody specifically detects PHLPP2 in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
| PHLPP2 | |
| Polyclonal | |
| Immunohistochemistry 1:50 - 1:200, Immunocytochemistry/ Immunofluorescence 0.25-2 ug/mL, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
| EC 3.1.3.16, KIAA0931PH domain leucine-rich repeat-containing protein phosphatase 2, PH domain and leucine rich repeat protein phosphatase 2, PH domain and leucine rich repeat protein phosphatase-like, PH domain leucine-rich repeat-containing protein phosphatase-like, PHLPPLPHLPP-like | |
| This antibody was developed against a recombinant protein corresponding to the amino acids: DLHLVLCTVETPASEICAGEGRESLYLQLHGDLVRRLEPTERPLQIVYDYLSRLGFDDPVRIQEEATNPDLGCMIRFYG | |
| 0.1 mL | |
| Cancer, Tumor Suppressors | |
| 23035 | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.2) and 40% Glycerol | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction