missing translation for 'onlineSavingsMsg'
Learn More

Phosphate as P @ 1000 μg/mL in H2O, VHG™

Product Code. 17980045
Change view
Click to view available options
Quantity:
100 mL
500 mL
Unit Size:
100mL
500mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17980045 500 mL 500mL
17999244 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17980045 Supplier VHG Labs Supplier No. VHGIPO4P500

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

VHG Labs offers ISO 17034 metallo-organic and aqueous inorganic certified reference materials for oil, fuel, and coolant analysis as well as a variety of certified standards for environmental, pharmaceutical, and industrial applications

Phosphate as P @ 1000 μg/mL in H2O
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Phosphate
CAS 14265-44-2
Methods Ion Chromatography/Wet Chemistry
Analyte or Component Names Phosphate
Shipping Condition Room Temperature
Molecular Formula (P O4)3-
Certifications/Compliance ISO 17034
Synonym PO43−
SMILES [O-]P(=O)([O-])[O-]
IUPAC Name Phosphate
Solvent or Matrix Water
Molecular Weight (g/mol) 94.97
Quantity 500 mL
Formula Weight 94.97 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.