missing translation for 'onlineSavingsMsg'
Learn More

Phosphate buffer solution, pH 4.5 EU Pharmacopoeia 0.05M, Fisher Chemical™
BIOPHARMA

Product Code. 12920684 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
1000 mL
Unit Size:
1000mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
12920684 1000 mL 1000mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12920684 Supplier Fisher Chemical Supplier No. SOL 038 1L
  • ONLINE EXCLUSIVE PRICE
    112.00 €
    110.44 € / 1000mL
    Save 1.56 €
    1% Off
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Pharmacopoeia reagents are intended for use in pharmacopoeia testing and monographs, produced under a GMP compliant ISO 9001 quality management system according to the relevant pharmacopoeia specification, using the pharmacopoeia reagent quality components and water meeting the relevant pharmacopoeia monograph requirement for Purified Water. Some pharmacopoeia standards are also accredited according to ISO 17025 and ISO Guide 43.

Specifications

CAS 7732-18-5
Concentration 0.05 M
pH 4.5
Quantity 1000 mL
Packaging Bottle
Grade Eu. Ph. Reference
Chemical Name or Material Phosphate buffer solution
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.