missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ Phosphate Buffered Saline

Product Code. 11454825 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
10 L
Unit Size:
10 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11454825 10 L 10 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11454825 Supplier MP Biomedicals™ Supplier No. 091760422

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Used in mammalian cell culture applications.

Phosphate Buffered Salts, Dulbecco's Formula (DPBS) is a balanced salt solution (BSS) used for the handling and culturing of mammalian cells. DPBS is used to irrigate, wash, and dilute mammalian cells. Phosphate buffering maintains the pH in the physiological range. Calcium and magnesium facilitate cell binding and clumping. DPBS without these ions can be used to wash and rinse suspended cells.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Phosphate Buffered Saline (PBS)
Quantity 10 L
Solution Type Phosphate Buffered Saline

Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.