missing translation for 'onlineSavingsMsg'
Learn More

Corning™ Phosphate Buffered Saline, 1X
BIOPHARMA

Product Code. 15313581
Change view
Click to view available options
Quantity:
6 x 500 mL
Unit Size:
Pack of 6
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15313581 6 x 500 mL Pack of 6
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15313581 Supplier Corning™ Supplier No. 21040CV
 Restricted Item
In Stock
Add to basket
Add to basket
This item is not returnable. View return policy

Used alone or in combination with other agents for washing tissues and cells.

  • Provide cells with water and inorganic ions
  • Maintains physiological pH and osmotic pressure
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Phosphate Buffered Saline (PBS)
Concentration 1 X
pH 7.4
Quantity 6 x 500 mL
Solution Type Phosphate Buffered Saline
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.